Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENTG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528410
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528410 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528410 Supplier Invitrogen™ Supplier No. PA528410
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GTPase-activating protein for the ADP ribosylation factor family (Potential). GTPase which may be involved in the degradation of expanded polyglutamine proteins through the ubiquitin-proteasome pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENTG3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 10% glycerol and 0.01% thimerosal; pH 7
Gene AGAP3
Gene Accession No. Q96P47
Gene Alias AGAP3; AGAP-3; ArfGAP with GTPase domain, ankyrin repeat and PH domain 3; Arf-GAP with GTPase, ANK repeat and PH domain-containing protein 3; centaurin; centaurin, gamma 3; centaurin-gamma-3; CENTG3; Cnt-g3; CRAG; CRAM-associated GTPase; CRMP (collapsin response mediator protein) associated; FLJ16146; FLJ34452; MR1-interacting protein; MRIP1; MRIP-1; MRIP-1 protein
Gene Symbols AGAP3
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human CENTG3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116988
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.