Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ CENTG3 Polyclonal Antibody
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553001
Description
Immunogen sequence: AKYEQKLFLA PLPSSDVPLG QQLLRAVVED DLRLLVMLLA HGSKEEVNET YGDGDGRTAL HLSSAMANVV FTQLLIWYGV DVRSRDARGL TPLAYARRAG SQECADILIQ HGCPGEGCGL APTPNREPAN GTNPSAE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
GTPase-activating protein for the ADP ribosylation factor family (Potential). GTPase which may be involved in the degradation of expanded polyglutamine proteins through the ubiquitin-proteasome pathway. 
            Specifications
| CENTG3 | |
| Polyclonal | |
| Unconjugated | |
| AGAP3 | |
| AGAP3; AGAP-3; ArfGAP with GTPase domain, ankyrin repeat and PH domain 3; Arf-GAP with GTPase, ANK repeat and PH domain-containing protein 3; centaurin; centaurin, gamma 3; centaurin-gamma-3; CENTG3; Cnt-g3; CRAG; CRAM-associated GTPase; CRMP (collapsin response mediator protein) associated; FLJ16146; FLJ34452; MR1-interacting protein; MRIP1; MRIP-1; MRIP-1 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116988 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | 
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96P47 | |
| AGAP3 | |
| Recombinant protein corresponding to Human CENTG3. Recombinant protein control fragment (Product #RP-102517). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
Safety and Handling
                                                    
                                                    WARNING: Cancer - www.P65Warnings.ca.gov 
                                                
                                            
                                        
                                    
                                
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                             
                             
                            