Learn More
CENTG3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302172AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
GTPase-activating protein for the ADP ribosylation factor family (Potential). GTPase which may be involved in the degradation of expanded polyglutamine proteins through the ubiquitin-proteasome pathway.Specifications
CENTG3 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q96P47 | |
AGAP3 | |
Between 825 and 875 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
AGAP3 | |
AGAP3; CENTG3; MRIP1; centaurin, gamma 3; MRIP-1; MRIP-1 protein; ArfGAP with GTPase domain, ankyrin repeat and PH domain 3; CRAG; FLJ16146; FLJ34452; CRMP (collapsin response mediator protein) associated; centaurin, gamma 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116988 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.