Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CENTG3 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518210
Description
This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
GTPase-activating protein for the ADP ribosylation factor family (Potential). GTPase which may be involved in the degradation of expanded polyglutamine proteins through the ubiquitin-proteasome pathway.
Specifications
| CENTG3 | |
| Polyclonal | |
| Unconjugated | |
| AGAP3 | |
| AGAP3; AGAP-3; ArfGAP with GTPase domain, ankyrin repeat and PH domain 3; Arf-GAP with GTPase, ANK repeat and PH domain-containing protein 3; centaurin; centaurin, gamma 3; centaurin-gamma-3; CENTG3; Cnt-g3; CRAG; CRAM-associated GTPase; CRMP (collapsin response mediator protein) associated; FLJ16146; FLJ34452; MR1-interacting protein; MRIP1; MRIP-1; MRIP-1 protein | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 116988 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q96P47 | |
| AGAP3 | |
| Synthetic peptide sequence (NPSAELHRSPSLL) corresponding to the C-terminus amino acids of AGAP3 (aa 863-875). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction