Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS™ Centrifuge Filters, Sterile

Catalog No. 09733116 Shop All Osmonics Products
Change view
Click to view available options
Volume (Metric):
1.5 mL
5 mL
Color:
Blue
Brown
Green
Pink
Rust
White
Material (Membrane):
Cellulose Acetate
Nitrocellulose
Nylon
Pore Size:
0.2 μm
0.45 μm
0.8 μm
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Material (Membrane) Pore Size
09-733-116 5 mL Rust Nitrocellulose 0.45 μm
09-733-100 1.5 mL Brown Nylon 0.2 μm
09-733-107 1.5 mL Blue Cellulose Acetate 0.2 μm
09-733-114 1.5 mL Pink Nitrocellulose 0.2 μm
09-733-110 1.5 mL White Cellulose Acetate 0.45 μm
09-733-115 1.5 mL Rust Nitrocellulose 0.45 μm
09-733-113 1.5 mL Green Cellulose Acetate 0.8 μm
09-733-101 5 mL Brown Nylon 0.2 μm
09-733-109 5 mL Blue Cellulose Acetate 0.2 μm
09-733-112 5 mL White Cellulose Acetate 0.45 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 09-733-116 Supplier GVS™ Supplier No. 10467019
Add to Cart
Add to Cart

Disposable filtration units are designed for centrifuge systems. Ideal for automated systems and high-speed batch filtration with robots

  • For the removal of particles or bacteria from aqueous or solvent solutions and gas streams
  • Membranes available: Cellulose Acetate, Nylon, NC(MCE)
  • Pore Sizes available: 0.2μm, 0.45μm, and 0.80μm
  • Rapid and simple preparation of a large number of samples
  • Considerably reduced contamination risk when working with radioactive biologically hazardous material
  • Application: 
    • Analytical Sample Prep, HPLC
    • Qualitative Analysis; Filtration of Aqueous Solutions
    • Rapid elution of agarose gels (0.45μm cellulose acetate)
    • Removal of particles and microorganisms from HPLC samples (nylon, cellulose acetate)
    • Rapid clearing and filtration of aqueous solutions (cellulose acetate, nitrocellulose)

Specifications

Autoclavable Autoclavable
Color Rust
Material Polypropylene
Material (Membrane) Nitrocellulose
Pore Size 0.45 μm
Sterility Sterile
No. per Pack 50
Quantity 50
Type Centrifuge Filter
Volume (Metric) 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.