Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Centrin 1 Antibody (OTI1E1), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270408AF532
Description
Centrin 1 Monoclonal specifically detects Centrin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Centrin 1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CETN1 | |
Full length recombinant protein of human Centrin 1 (NP_004057) produced in E.coli. | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI1E1 | |
Western Blot, Immunohistochemistry | |
calcium binding protein, caltractin, Caltractin isoform 2, CEN1centrin-1, centrin, EF-hand protein, 1, CETN, EF-hand protein | |
Mouse | |
0.1 mL | |
1068 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction