Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Centrin 1 Antibody (OTI1E1), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270408B
Description
Centrin 1 Monoclonal specifically detects Centrin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Centrin 1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| CETN1 | |
| Full length recombinant protein of human Centrin 1 (NP_004057) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI1E1 | |
| Western Blot, Immunohistochemistry | |
| calcium binding protein, caltractin, Caltractin isoform 2, CEN1centrin-1, centrin, EF-hand protein, 1, CETN, EF-hand protein | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 1068.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction