Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Centrin 1 Antibody (OTI1E1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246253
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
Centrin 1 Monoclonal specifically detects Centrin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Centrin 1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
calcium binding protein, caltractin, Caltractin isoform 2, CEN1centrin-1, centrin, EF-hand protein, 1, CETN, EF-hand protein | |
Mouse | |
19.4 kDa | |
0.1 mL | |
Cell Cycle and Replication | |
1068 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI1E1 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
CETN1 | |
Full length human recombinant protein of human CETN1 (NP_004057) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction