Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Centrin 1 Monoclonal Antibody (GT147)
GREENER_CHOICE

Catalog No. PIMA531517
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531517 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531517 Supplier Invitrogen™ Supplier No. MA531517
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Keep as concentrated solution. Positive Control: A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene plays important roles in the determination of centrosome position and segregation, and in the process of microtubule severing. This protein is localized to the centrosome of interphase cells, and redistributes to the region of the spindle poles during mitosis, reflecting the dynamic behavior of the centrosome during the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Centrin 1
Applications Western Blot
Classification Monoclonal
Clone GT147
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CETN1
Gene Accession No. Q12798
Gene Alias calcium binding protein; Calt; Caltractin; Caltractin isoform 2; CEN1; centrin 1; centrin 1 S homeolog; centrin, EF-hand protein, 1; centrin, EF-hand protein, 2; centrin-1; CETN; cetn1; cetn1.S; cetn2; EF-hand protein; testicular tissue protein Li 37; testis-specific murine centrin; Unknown (protein for MGC:133570); xcen; XELAEV_18042289mg
Gene Symbols CETN1
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Centrin 1. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1068
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.