Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Centrin 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104315
Description
Antibody detects endogenous levels of total Centrin 1.
The protein encoded by this gene plays important roles in the determination of centrosome position and segregation, and in the process of microtubule severing. This protein is localized to the centrosome of interphase cells, and redistributes to the region of the spindle poles during mitosis, reflecting the dynamic behavior of the centrosome during the cell cycle.
Specifications
Centrin 1 | |
Polyclonal | |
Unconjugated | |
CETN1 | |
calcium binding protein; Calt; Caltractin; Caltractin isoform 2; CEN1; centrin 1; centrin 1 S homeolog; centrin, EF-hand protein, 1; centrin, EF-hand protein, 2; centrin-1; CETN; cetn1; cetn1.S; cetn2; EF-hand protein; testicular tissue protein Li 37; testis-specific murine centrin; Unknown (protein for MGC:133570); xcen; XELAEV_18042289mg | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1068, 26369, 84592 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P41209, Q12798 | |
CETN1 | |
A synthesized peptide derived from human CETN1(Accession Q12798), corresponding to amino acid residues T94-E144. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction