Learn More
Invitrogen™ Centrin 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592443
Description
Immunogen sequence: MASNFKKANM ASSSQRKRMS PKPELTEEQK QEIREAFDLF DADGTGTIDV KELKVAMRAL GFEPKKEEIK KMISEIDKEG TGKMNFGDFL TVMTQKMSEK DTKEEILKAF KLFDDDETGK ISFKNLKRVA KELGENLTDE ELQEMIDEAD RDGDGEVSEQ EFLRIMKKTS LY; Positive Samples: U-251MG, SKOV3, HeLa, HepG2, A-549, Mouse brain, Mouse kidney, Rat brain; Cellular Location: Cytoplasm, Nucleus, centriole, centrosome, cytoskeleton, microtubule organizing center.
RNP2 a type I integral membrane protein found only in the rough endoplasmic reticulum. The encoded protein is part of an N-oligosaccharyl transferase complex that links high mannose oligosaccharides to asparagine residues found in the Asn-X-Ser/Thr consensus motif of nascent polypeptide chains. This protein is similar in sequence to the yeast oligosaccharyl transferase subunit SWP1.
Specifications
Centrin 2 | |
Polyclonal | |
Unconjugated | |
CETN2 | |
1110034A02Rik; AI326150; CALT; Caltractin; caltractin (20kD calcium-binding protein); Caltractin isoform 1; caltractin, 20 kDa calcium binding protein; CEN2; centrin 2; centrin, EF-hand protein, 2; Centrin2; centrin-2; Cetn2; EF hand protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1069, 26370, 84593 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
2.23 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P41208, Q9R1K9 | |
CETN2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-172 of human CETN2 (NP_0043351). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.