Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Centrin 3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Centrin 3 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Centrin 3 Polyclonal specifically detects Centrin 3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Centrin 3 | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication | |
CDC31 yeast homolog, CEN3centrin, EF-hand protein, 3 (CDC31 homolog, yeast), centrin, EF-hand protein, 3, centrin, EF-hand protein, 3 (CDC31 yeast homolog), centrin-3, EF-hand superfamily member, MGC12502, MGC138245 | |
CETN3 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
1070 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MSLALRSELVVDKTKRKKRRELSEEQ | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title