Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Centrin 3 Antibody (OTI4G5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270409H
Description
Centrin 3 Monoclonal specifically detects Centrin 3 in Human, Mouse samples. It is validated for Western Blot.Specifications
Centrin 3 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CETN3 | |
Full length recombinant protein of human Centrin 3 (NP_004356) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 |
Western Blot | |
OTI4G5 | |
Western Blot | |
CDC31 yeast homolog, CEN3centrin, EF-hand protein, 3 (CDC31 homolog, yeast), centrin, EF-hand protein, 3, centrin, EF-hand protein, 3 (CDC31 yeast homolog), centrin-3, EF-hand superfamily member, MGC12502, MGC138245 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
1070.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction