Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Centrin 3 Antibody (OTI4G5), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270409PCP
Description
Centrin 3 Monoclonal antibody specifically detects Centrin 3 in Human, Mouse samples. It is validated for Western BlotSpecifications
Centrin 3 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
OTI4G5 | |
Western Blot | |
CDC31 yeast homolog, CEN3centrin, EF-hand protein, 3 (CDC31 homolog, yeast), centrin, EF-hand protein, 3, centrin, EF-hand protein, 3 (CDC31 yeast homolog), centrin-3, EF-hand superfamily member, MGC12502, MGC138245 | |
Full length human recombinant protein of human CETN3 (NP_004356) produced in E.coli. | |
0.1 mL | |
Cell Cycle and Replication | |
1070 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction