Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Centrin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557337
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557337 Supplier Invitrogen™ Supplier No. PA557337
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AMRALGFDVK KADVLKILKD YDREATGKIT FEDFNEVVTD WILERDPHEE ILK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The protein encoded by this gene contains four EF-hand calcium binding domains, and is a member of the centrin protein family. Centrins are evolutionarily conserved proteins similar to the CDC31 protein of S. cerevisiae. Yeast CDC31 is located at the centrosome of interphase and mitotic cells, where it plays a fundamental role in centrosome duplication and separation. Multiple forms of the proteins similar to the yeast centrin have been identified in human and other mammalian cells, some of which have been shown to be associated with centrosome fractions. This protein appears to be one of the most abundant centrins associated with centrosome, which suggests a similar function to its yeast counterpart. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Centrin 3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cetn3
Gene Accession No. O15182
Gene Alias CDC31; CDC31 yeast homolog; CEN3; centrin 3; centrin, EF-hand protein, 3; centrin, EF-hand protein, 3 (CDC31 homolog, yeast); centrin-3; Cetn3; EF-hand superfamily member; MmCEN3
Gene Symbols Cetn3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Centrin 3. Recombinant protein control fragment (Product #RP-96905).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1070
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.