Learn More
Description
High affinity and selective PLK4 inhibitor (Ki = 0.16 nM). Exhibits >1000-fold selectivity for PLK4 over Aurora A and Aurora B. Depletes centriole and centrosome levels in vitro. Induces cell cycle arrest in normal human cell lines in a p53-dependent manner. Decreases expression of pluripotency markers and induces differentiation in pluripotent stem cells.
Specifications
Specifications
| Chemical Name or Material | 2-[[2-Fluoro-4-[[(2-fluoro-3-nitrophenyl)methyl]sulfonyl]phenyl]thio]-5-methoxy-N-(5-methyl-1H-pyrazol-3-yl)-6-(4-morpholinyl)-4-pyrimidinamine |
| CAS | 1798871-30-3 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Target | Polo-like Kinase Inhibitors |
| Molecular Formula | C26H25F2N7O6S2 |
| Formula Weight | 633.65 |
| Purity | >98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

