Learn More
Centrisart™ I centrifugal ultrafiltration unit, Sold by MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Pore Size | 100 kDa MWCO |
| Pore Volume | 2.5 mL |
| Quantity | 12 Tubes |
| Surface Area | 0.79 cm2 |
| Volume (Metric) | 15 mL |
| Material | Polyethersulfon |
Centrisart is a registered trademark of Sartorius Stedim Biotech GmbH
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.