Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Centromeric histone H3-like protein-2 Rabbit anti-Plant, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS0621R

Description
Centromeric histone H3-like protein-2. Also involved in antibodies against CENPA are present in sera from patients with autoimmune diseases that developed autoantibodies against centrosomal proteins.Specifications
Centromeric histone H3-like protein-2 | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Protein A | |
RUO | |
Plant | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
KLH conjugated synthetic peptide derived from Brassica juncea Histone H3-like protein. | |
100 μL | |
Primary | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction