Learn More
Brady Centurion™ Flood Barriers
Catalog No. p-200115235
Click to view available options
Length (English):
5 ft.
10 ft.
17 ft.
Enjoy rapid response and none of the labor or mess of sandbags with water-activated Centurion™ Flood Barriers.
Transform flood readiness. Maximize asset protection. Enjoy rapid response and none of the labor or mess of sandbags with water-activated Centurion™ Flood Barriers. Freshwater-swelled barriers contain, control and divert flood water. Absorbent-filled design swells to full size on contact with water in 10 minutes. Flat and lightweight when dry for space-saving storage and easy positioning before first use. Easier and cleaner than sandbags. Built-in wedge prevents rolling. To divert flowing water, offset each downstream barrier toward dry side and slightly behind upstream barrier. To stack barriers, place wedge on top of wedge with two barriers, then place a third barrier on top in the middle. Cannot be used with salt water.
  • Freshwater-swelled barriers contain, control and divert flood water
  • Absorbent-filled design swells to full size on contact with water in 10 minutes
  • Flat and lightweight when dry for space-saving storage and easy positioning before first use
Specifications
Material Polypropylene
Type Spill Control System
Packaging Quantity 10/Cs.
Length (English) 10 ft.
Diameter (English) 6 in.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.