Learn More
3M™ Centurion™ Splash Goggles
Catalog No. p-2908965 Shop All 3M Products
Click to view available options
Lens Coating:
Anti-fog
Uncoated
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.
Open viewing range promotes compliance
  • Wide-angle wraparound design offers exceptional side-to-side peripheral vision
  • Polycarbonate lens absorbs 99.9% UV
  • Large lens opening to accommodate prescription and safety eyewear
  • Wide nasal flare
  • Vertical, indirect-venting system to circulate air over lens
  • 3M Catalog# 40304-00000-10 , 3M Catalog# 40305-00000-10
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3-2007 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Tint Clear
Product Type Splash Goggles
Model 40305-00000-10
Material Polycarbonate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.