Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP104/KIAA0562 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981862
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1862 Supplier Bioss Supplier No. BS12005R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ionotropic glutamate receptors are oligomeric ion channel proteins, which mediate fast neurotransmission and excitatory synapses in the mammalian central nervous system. There are three distinct subfamilies, of which NMDA receptors have received particular attention because they are implicated in both synaptic plasticity and the pathogenesis of acute and chronic neurodegenerative disorders. A unique feature of the NMDA receptors is their requirement for both glutamate and the co-agonist glycine for efficient gating. Glycine increases the affinity of the receptor for the endogenous agonist glutamate. The glycine binding site is found on the NR1 subunit of the NMDA receptor.

Specifications

Antigen CEP104/KIAA0562
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CEP104
Gene Accession No. O60308
Gene Alias centrosomal protein 104; centrosomal protein 104kDa; Centrosomal protein of 104 kDa; CEP104; CFAP256; GlyBP; JBTS25; KIAA0562; ROC22
Gene Symbols CEP104
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CEP104/KIAA0562.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9731
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.