Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP112 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555088 Supplier Invitrogen™ Supplier No. PA555088
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RNTLHKEKDH LVNDYEQNMK LLQTKYDADI NLLKQEHALS ASKASSMIEE LEQNVCQLKQ QLQESELQRK QQLRDQE Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 77%.

CCDC46 is a protein with filament, myosin tail and ATPase domains. Orthologs of the gene exist in mouse, rat and chimp.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP112
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEP112
Gene Accession No. Q8N8E3
Gene Alias 1700001M19Rik; 1700029K01Rik; 8430407H02Rik; AV043680; AV207351; CCDC46; centrosomal protein 112; centrosomal protein 112kDa; centrosomal protein of 112 kDa; centrosomal protein of 112 kDa; LOW QUALITY PROTEIN: centrosomal protein of 112 kDa; Cep112; coiled-coil domain containing 46; coiled-coil domain-containing protein 46; Macoco; RGD1564168
Gene Symbols CEP112
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CEP112. Recombinant protein control fragment (Product #RP-93358).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201134
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.