Learn More
Invitrogen™ CEP112 Polyclonal Antibody

Description
Immunogen sequence: LMPASLRQEL EDTISSLKSQ VNFLQKRASI LQEELTTY Antigen sequence identity: Mouse 100%; Rat 100%.
Specifications
Specifications
| Antigen | CEP112 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CEP112 |
| Gene Accession No. | Q8N8E3 |
| Gene Alias | 1700001M19Rik; 1700029K01Rik; 8430407H02Rik; AV043680; AV207351; CCDC46; centrosomal protein 112; centrosomal protein 112kDa; centrosomal protein of 112 kDa; centrosomal protein of 112 kDa; LOW QUALITY PROTEIN: centrosomal protein of 112 kDa; Cep112; coiled-coil domain containing 46; coiled-coil domain-containing protein 46; Macoco; RGD1564168 |
| Gene Symbols | CEP112 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.