Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP120 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109513
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109513 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109513 Supplier Invitrogen™ Supplier No. PA5109513
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVSKSDQLLI VVSILEGRHF PKRPKHMLVV EAKFDGEQLA TDPVDHTDQP EFATELAWEI DRKALHQHRL QRTPIKLQCF ALDPVTSAKE TIGYIVLDLR TAQETKQAPK WYQLLSNKYT KFKSEIQISI ALETDTKPPV DSFKAKGAPP RDGKVPAILA GLDPRDIVAV LNEEGGYHQI GPAEYCTDSF IMSVTIAFAT QLEQLIPCTM KLPERQPEFF FYYSLLGNDV TNEPFNDLIN PNFEPERASV RIRSSVEILR VYLALQSKLQ.

This gene encodes a protein that functions in the microtubule-dependent coupling of the nucleus and the centrosome. A similar protein in mouse plays a role in both interkinetic nuclear migration, which is a characteristic pattern of nuclear movement in neural progenitors, and in neural progenitor self-renewal. Mutations in this gene are predicted to result in neurogenic defects. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP120
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CEP120
Gene Accession No. Q7TSG1, Q8N960
Gene Alias A230075C01; AU016693; Ccdc100; centrosomal protein 120; centrosomal protein 120kDa; centrosomal protein of 120 kDa; Cep120; coiled-coil domain containing 100; coiled-coil domain-containing protein 100; RGD1565619; SRTD13
Gene Symbols CEP120
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human CEP120 (NP_694955.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153241, 225523, 307302
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.