Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP128 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178745JF549
Description
CEP128 Polyclonal antibody specifically detects CEP128 in Human samples. It is validated for Western BlotSpecifications
CEP128 | |
Polyclonal | |
Western Blot | |
centrosomal protein 128kDa | |
The immunogen for this product maps to a region between residue 1044 and 1094 of human Centrosomal Protein 128kDa using the numbering given in entry NP_689659.2 (GeneID 145508). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
145508 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction