Learn More
CEP128 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303348A

Description
The recommended shelf life for this product is 1 year from date of receipt.
CEP128 gene ontology annotations related to this gene include protein localization.Specifications
CEP128 | |
Polyclonal | |
Unconjugated | |
CEP128 | |
Centrosomal protein of 128 kDa, Cep128, CEP128, C14orf145, C14orf61 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
145508 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q6ZU80 | |
CEP128 | |
Between 1044 and 1094 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.