Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP131 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554953
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554953 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554953 Supplier Invitrogen™ Supplier No. PA554953
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DFLMLFEGSP SGKKRPASLS TAPSEKGATW NVLDDQPRGF TLPSNARSSS ALDSPAGPRR KECTVALAPN FTANNRSNKG AVGNCVTTM Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 75%.

AZI1, also named as CEP131 and KIAA1118, may play a role in spermatogenesis. It is located at the ciliary base and required for cilium formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP131
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEP131
Gene Accession No. Q9UPN4
Gene Alias 5-azacytidine induced 1; 5-azacytidine induced gene 1; 5-azacytidine-induced protein 1; Az1; Azi; AZI1; centrosomal protein 131; centrosomal protein 131 kDa; centrosomal protein 131kDa; centrosomal protein of 131 kDa; CEP131; KIAA1118; Pre-acrosome localization protein 1; ZA1
Gene Symbols CEP131
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CEP131. Recombinant protein control fragment (Product #RP-92302).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22994
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.