Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEP131 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117862
This item is not returnable.
View return policy
Description
CEP131 Polyclonal Antibody for Western Blot, IP
AZI1, also named as CEP131 and KIAA1118, may play a role in spermatogenesis. It is located at the ciliary base and required for cilium formation.
Specifications
CEP131 | |
Polyclonal | |
PBS with 0.03% ProClin 300; pH 7 | |
Q9UPN4 | |
CEP131 | |
E. coli-derived Human CEP131/AZI1 fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
CEP131 | |
5-azacytidine induced 1; 5-azacytidine induced gene 1; 5-azacytidine-induced protein 1; Az1; Azi; AZI1; centrosomal protein 131; centrosomal protein 131 kDa; centrosomal protein 131kDa; centrosomal protein of 131 kDa; CEP131; KIAA1118; Pre-acrosome localization protein 1; ZA1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
22994 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction