Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP135 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539089
Description
This gene encodes a centrosomal protein, which acts as a scaffolding protein during early centriole biogenesis, and is also required for centriole-centriole cohesion during interphase. Mutations in this gene are associated with autosomal recessive primary microcephaly-8.
Specifications
CEP135 | |
Polyclonal | |
Unconjugated | |
CEP135 | |
centrosomal protein 135 kDa, centrosomal protein 135kDa, Centrosomal protein 4, Centrosomal protein of 135 kDa, centrosome protein 4, centrosome protein cep135, Cep135, Cep135, CEP4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
381644, 9662 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q66GS9, Q6P5D4 | |
CEP135 | |
A synthetic peptide derived from the C-terminal region of human CEP135 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction