Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP135 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE1857MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1857MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1857MI Supplier MilliporeSigma Supplier No. ABE1857
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-CEP135 is a highly specific rabbit polyclonal antibody that targets Centrosomal protein of 135 kDa and has been tested in Electron Microscopy, Immunocytochemistry, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP135
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CEP135,CEP4,KIAA0635
Host Species Rabbit
Immunogen His-tagged recombinant fragment corresponding to 492 amino acids from the C-termnal region of human Cep135.
Purification Method Protein A Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_079285
Test Specificity This polyclonal antibody detects centrosomal protein of 135 kDa in human cells. It targets an epitope within 492 amino acids from the C-terminal half.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.