Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP135 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310683100UL
Description
CEP135 Polyclonal specifically detects CEP135 in Human samples. It is validated for Western Blot.Specifications
| CEP135 | |
| Polyclonal | |
| Western Blot 1.0 ug/ml | |
| centrosomal protein 135kDa, Centrosomal protein 4centrosomal protein of 135 kDa, Cep135, CEP4, FLJ13621, KIAA0635centrosome protein cep135 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human CEP135 (NP_079285). Peptide sequence REHSDQHVKELKTSLKKCARETADLKFLNNQYAHKLKLLEKESKAKNERI | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9662 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction