Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP152 Antibody (1315), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP243721
Description
Ensure accurate, reproducible results in Western Blot
CEP152 Monoclonal specifically detects CEP152 in Human samples. It is validated for Western Blot.Specifications
CEP152 | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
CEP152 | |
Recombinant protein encompassing a sequence within the N-terminus region of human CEP152. The exact sequence is proprietary. | |
Protein G purified | |
RUO | |
22995 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot | |
1315 | |
Western Blot 1:500-1:3000 | |
centrosomal protein 152kDa, centrosomal protein of 152 kDa, Cep152, FLJ21594, KIAA0912MCPH4 | |
Mouse | |
189 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction