Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP152 Monoclonal Antibody (P1285)

Catalog No. PIMA518285
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518285 Supplier Invitrogen™ Supplier No. MA518285
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the cytoskeletal BTB/kelch repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP152
Applications Western Blot
Classification Monoclonal
Clone P1285
Concentration 1.65 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CEP152
Gene Accession No. O94986
Gene Alias AI851464; asterless; CC-CKR-8; CCR-8; CDw198; centrosomal protein 152; centrosomal protein 152kDa; centrosomal protein of 152 kDa; CEP152; CKRL1; CMKBR8; CMKBRL2; CY6; GPRCY6; Kiaa0912; MCPH4; MCPH9; microcephaly, primary autosomal recessive 4; mKIAA0912; RGD1305348; SCKL5; TER1
Gene Symbols CEP152
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human CEP152. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22995
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.