Learn More
Description
Specifications
Specifications
| Antigen | Cep152 (P7) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | Please refer to lot specific datasheet. |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal antibody in buffer containing 80mM Tris (pH 8.0), 40mM glycine, and 50% ethylene glycol with 0.05% Sodium Azide. |
| Gene | CEP152, KIAA0912, MCPH9, SCKL5 |
| Gene Accession No. | O94986 |
| Host Species | Rabbit |
| Immunogen | Recombinant human Cep152 C-terminal fragment. |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.