Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP152 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE1856MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1856MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1856MI Supplier MilliporeSigma Supplier No. ABE1856
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-CEP152, N-Term is a highly specific rabbit polyclonal antibody that targets Centrosomal protein of 152KDa and has been tested in Electron Microscopy, Immunocytochemistry, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP152
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Cep152,KIAA0912
Host Species Rabbit
Immunogen His-tagged recombinat fragment corresponding to 87 amino acids from the N-terminal end of human Centrosomal protein of 152 kDa (CEP152).
Purification Method Protein A Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_055800
Test Specificity This polyclonal antibody specifically detects human CEP152 protein. It targets an epitope within the first 87 amino acids from the N-terminal region. Recognizes both long and short isoforms of Cep152.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.