Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEP164 Monoclonal Antibody (13)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527823
Description
Positive Control: HeLa Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CEP164 was initially identified as a centrosomal protein, but other studies have indicated that it also plays a role in the formation of primary cilia, the microtubule-based sensory antennae projecting from the surface of many eukaryotic cells as well as in DNA damage response acting as a mediator protein. CEP164 interacts with both ATR and ATM, proteins that trigger a number of cellular responses including the initiation of DNA damaged-induced cell cycle checkpoints. It is phosphorylated upon replication stress, ultraviolet (UV) radiation, and ionizing radiation silencing of CEP164 significantly reduces the DNA damage-induced phosphorylation of several proteins in the DNA damage-activated signaling cascade and compromises cell survival after UV damage. At least two isoforms of CEP164 are known to exist.
Specifications
CEP164 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9UPV0 | |
CEP164 | |
Human CEP164. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
13 | |
Unconjugated | |
CEP164 | |
AI450905; BC027092; ce164; centrosomal protein 164; centrosomal protein 164kDa; centrosomal protein of 164 kDa; Cep 164; CEP164; D030051D21; KIAA1052; mKIAA1052; NPHP15; RGD1560988; RGD1561243 | |
Mouse | |
Protein G | |
RUO | |
22897 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction