Learn More
Invitrogen™ CEP164 Polyclonal Antibody

Description
Immunogen sequence: EVRSTEPVAP PEQLSEAALK AMEEAVAQVL EQDQRHLLES KQEKMQQLRE KLCQEEEEEI LRLHQQKEQS LSSLRERLQK AIEEE Highest antigen sequence indentity to the following orthologs: Mouse - 40%, Rat - 61%.
Specifications
Specifications
| Antigen | CEP164 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CEP164 |
| Gene Accession No. | Q9UPV0 |
| Gene Alias | AI450905; BC027092; ce164; centrosomal protein 164; centrosomal protein 164kDa; centrosomal protein of 164 kDa; Cep 164; CEP164; D030051D21; KIAA1052; mKIAA1052; NPHP15; RGD1560988; RGD1561243 |
| Gene Symbols | CEP164 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.