Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP170 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP26866425UL
Description
CEP170 Polyclonal antibody specifically detects CEP170 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
CEP170 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
centrosomal protein 170kDa, Cep170, KARP 1 binding protein, KARP-1-binding protein, KARP1-binding protein, KIAA0470FAM68AKABcentrosomal protein of 170 kDa, XRCC5 binding protein | |
This antibody was developed against a recombinant protein corresponding to amino acids: KQLQAINAMIDPDGTLEALNNMGFPSAMLPSPPKQKSSPVNNHHSPGQTPTL | |
25 μL | |
Cell Biology, Cell Cycle and Replication | |
9859 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction