Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP170 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111221
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111221 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111221 Supplier Invitrogen™ Supplier No. PA5111221
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQWASLAANH TRHDQEERIM EFSAPLPLEN ETEISESGMT VRSTGSATSL ASQGERRRRT LPQLPNEEKS LESHRAKVVT QRSEIGEKQD TELQEKETP Highest antigen sequence indentity to the following orthologs: Mouse - 76%, Rat - 75%.

The product of this gene is a component of the centrosome, a non-membraneous organelle that functions as the major microtubule-organizing center in animal cells. During interphase, the encoded protein localizes to the sub-distal appendages of mature centrioles, which are microtubule-based structures thought to help organize centrosomes. During mitosis, the protein associates with spindle microtubules near the centrosomes. The protein interacts with and is phosphorylated by polo-like kinase 1, and functions in maintaining microtubule organization and cell morphology. The human genome contains a putative transcribed pseudogene. Several alternatively spliced transcript variants of this gene have been found, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP170
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEP170
Gene Accession No. Q5SW79
Gene Alias 4933426L22Rik; A330004A13Rik; AI195353; centrosomal protein 170; centrosomal protein 170kDa; centrosomal protein of 170 kDa; CEP170; FAM68A; KAB; KARP-1-binding protein; KARP1-binding protein; KIAA0470; XRCC5 binding protein
Gene Symbols CEP170
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human CEP170. Recombinant protein control fragment (Product #RP-94301).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9859
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.