Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEP170 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583782
Description
Immunogen sequence: KQLQAINAMI DPDGTLEALN NMGFPSAMLP SPPKQKSSPV NNHHSPGQTP TL.
The product of this gene is a component of the centrosome, a non-membraneous organelle that functions as the major microtubule-organizing center in animal cells. During interphase, the encoded protein localizes to the sub-distal appendages of mature centrioles, which are microtubule-based structures thought to help organize centrosomes. During mitosis, the protein associates with spindle microtubules near the centrosomes. The protein interacts with and is phosphorylated by polo-like kinase 1, and functions in maintaining microtubule organization and cell morphology. The human genome contains a putative transcribed pseudogene. Several alternatively spliced transcript variants of this gene have been found, but the full-length nature of some of these variants has not been determined.
Specifications
CEP170 | |
Polyclonal | |
Unconjugated | |
CEP170 | |
4933426L22Rik; A330004A13Rik; AI195353; centrosomal protein 170; centrosomal protein 170kDa; centrosomal protein of 170 kDa; CEP170; FAM68A; KAB; KARP-1-binding protein; KARP1-binding protein; KIAA0470; XRCC5 binding protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9859 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5SW79 | |
CEP170 | |
Recombinant protein corresponding to Human CEP170. Recombinant protein control fragment (Product #RP-108368). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction