Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEP170B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563645
Description
Immunogen sequence: QPLRAQKEMS PSPPAAQDPG GTALVSAREQ SSERQHHPLG PTDMGRGEPV RRSAIRRGHR PRGSLDWPSE ERGPVLAHLP SSDVMASNHE T Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 58%.
CEP170B plays a role in microtubule organization.
Specifications
CEP170B | |
Polyclonal | |
Unconjugated | |
CEP170B | |
AI466840; AW555464; Centrosomal protein 170B; centrosomal protein of 170 kDa protein B; CEP170B; CEP170R; Cep170-related; FAM68C; KIAA0284; mKIAA0284; si:ch73-160l8.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
283638 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y4F5 | |
CEP170B | |
Recombinant protein corresponding to Human CEP170B. Recombinant protein control fragment (Product #RP-101922). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction