Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEP19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561556
Description
Immunogen sequence: MCTAKKCGIR FQPPAIILIY ESEIKGKIRQ RIMPVRNFSK FSDCTRAAEQ LKNNPRHKSY LEQVSLRQLE KLFS Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.
The protein encoded by this gene localizes to centrosomes and primary cilia and co-localizes with a marker for the mother centriole. This gene resides in a region of human chromosome 3 that is linked to morbid obesity. A homozygous knockout of the orthologous gene in mouse resulted in mice with morbid obesity, hyperphagy, glucose intolerance, and insulin resistance. Mutations in this gene cause morbid obesity and spermatogenic failure (MOSPGF). This gene has a pseudogene on human chromosome 2.
Specifications
CEP19 | |
Polyclonal | |
Unconjugated | |
CEP19 | |
1500031L02Rik; AI428934; AL022620; C3orf34; centrosomal protein 19; centrosomal protein 19kDa; centrosomal protein of 19 kDa; Cep19; HSD5; MOSPGF; RGD621352; testis specific protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84984 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96LK0 | |
CEP19 | |
Recombinant protein corresponding to Human CEP19. Recombinant protein control fragment (Product #RP-98223). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction