Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP192 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128718V
Description
CEP192 Polyclonal antibody specifically detects CEP192 in Human samples. It is validated for Western BlotSpecifications
CEP192 | |
Polyclonal | |
Western Blot | |
centrosomal protein 192kDa, centrosomal protein of 192 kDa, Cep192, FLJ10352, KIAA1569 | |
The immunogen recognized by this antibody maps to a region between residue 1800 and 1850 of human centrosomal protein using the numbering given in entry NP_115518.2 (GeneID 55125). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55125 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction