Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP192 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559199
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559199 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559199 Supplier Invitrogen™ Supplier No. PA559199
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FVLKERTQEN VTLIYNPSDR GINNKTATEL STVYLFGGDE ISRQQYRRAL LHKPEMIKQI LPEHSVLQNI NFVEAFQDEL LVTEVYD Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 74%.

CEP192 is required for mitotic centrosome and spindle assembly. Appears to be a major regulator of pericentriolar material (PCM) recruitment, centrosome maturation, and centriole duplication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP192
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEP192
Gene Accession No. Q8TEP8
Gene Alias 192 kDa centrosomal protein; centrosomal protein 192; centrosomal protein 192kDa; centrosomal protein of 192 kDa; CEP192; FLJ10352; KIAA1569; PP8407; PPP1R62; protein phosphatase 1, regulatory subunit 62
Gene Symbols CEP192
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CEP192. Recombinant protein control fragment (Product #RP-98208).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55125
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.