Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEP250 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568141
Description
Predicted to react with Human samples.
Probably plays an important role in centrosome cohesion during interphase.
Specifications
CEP250 | |
Polyclonal | |
Unconjugated | |
CEP250 | |
250 kDa centrosomal protein; AW490617; B230210E21Rik; centrosomal Nek2-associated protein 1; Centrosomal protein 2; centrosomal protein 250; centrosomal protein 250kDa; centrosome associated protein; centrosome-associated protein CEP250; Cep2; CEP250; CNAP1; C-Nap1; Inmp; Intranuclear matrix protein | |
Rabbit | |
affinity chromatography | |
RUO | |
11190 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9BV73 | |
CEP250 | |
Synthesized peptide derived from human CEP250. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction