Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP250 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143966
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143966 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143966 Supplier Invitrogen™ Supplier No. PA5143966
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human 293T whole cell, human HEL whole cell. IHC: human placenta tissue, human spleen tissue, human liver cancer tissue, mouse brain tissue, rat brain tissue. Flow: SiHa cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Probably plays an important role in centrosome cohesion during interphase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP250
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene CEP250
Gene Accession No. Q60952, Q9BV73
Gene Alias 250 kDa centrosomal protein; AW490617; B230210E21Rik; centrosomal Nek2-associated protein 1; Centrosomal protein 2; centrosomal protein 250; centrosomal protein 250kDa; centrosome associated protein; centrosome-associated protein CEP250; Cep2; CEP250; CNAP1; C-Nap1; Inmp; Intranuclear matrix protein
Gene Symbols CEP250
Host Species Rabbit
Immunogen E.coli-derived human CEP250 recombinant protein (Position: Q563-K853).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11190, 16328, 311573
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.