Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP250 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566730 Supplier Invitrogen™ Supplier No. PA566730
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HSPGATSTAEL GSRGEQGVQL GEVSGVEAEP SPDGMEKQSW RQRLEHLQQA VARLEIDRSR LQRHNVQLRS TLEQVERERR Highest antigen sequence identity to the following orthologs - mouse 60%, rat 60%.

Probably plays an important role in centrosome cohesion during interphase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP250
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEP250
Gene Accession No. Q9BV73
Gene Alias 250 kDa centrosomal protein; AW490617; B230210E21Rik; centrosomal Nek2-associated protein 1; Centrosomal protein 2; centrosomal protein 250; centrosomal protein 250kDa; centrosome associated protein; centrosome-associated protein CEP250; Cep2; CEP250; CNAP1; C-Nap1; Inmp; Intranuclear matrix protein
Gene Symbols CEP250
Host Species Rabbit
Immunogen Recombinant Human CEP250. Recombinant protein control fragment (Product #RP-107382).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11190
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.