Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP27 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215860
Description
CEP27 Polyclonal specifically detects CEP27 in Human samples. It is validated for Western Blot.Specifications
CEP27 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
centrosomal protein 27kDa, Centrosomal protein of 27 kDa, CEP27C15orf25, chromosome 15 open reading frame 25, FLJ10460, HAUS augmin-like complex subunit 2, HAUS augmin-like complex, subunit 2, HsT17025 | |
Rabbit | |
27 kDa | |
0.1 mL | |
Primary | |
Human. Expected to cross react based on sequence identity: Monkey (81%), Bovine (87%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
1.02mg/mL | |
Western Blot 1:500-1:3000 | |
Q9NVX0 | |
HAUS2 | |
Recombinant protein encompassing a sequence within the center region of human HAUS2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
55142 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction