Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP290 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10086991AF700
Description
CEP290 Polyclonal specifically detects CEP290 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CEP290 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
BBS14Bardet-Biedl syndrome 14 protein, Cancer/testis antigen 87, centrosomal protein 290kDa, Cep290, CT87JBTS6, FLJ13615, JBTS5CTCL tumor antigen se2-2, KIAA0373FLJ21979, LCA10monoclonal 3H11 antigen, MKS4, Nephrocystin-6, NPHP6centrosomal protein of 290 kDa, POC3, POC3 centriolar protein homolog, prostate cancer antigen T21, rd16,3H11AG, SLSN6nephrocytsin-6,3H11Ag, Tumor antigen se2-2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CEP290 | |
The immunogen recognized by this antibody maps to a region between residue 2429 and 2479 of human centrosomal protein 290kDa using the numbering given in entry NP_079390.3 (GeneID 80184). | |
100 μL | |
Vision | |
80184 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction