Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP290 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25710425UL
Description
CEP290 Polyclonal specifically detects CEP290 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
CEP290 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
BBS14Bardet-Biedl syndrome 14 protein, Cancer/testis antigen 87, centrosomal protein 290kDa, Cep290, CT87JBTS6, FLJ13615, JBTS5CTCL tumor antigen se2-2, KIAA0373FLJ21979, LCA10monoclonal 3H11 antigen, MKS4, Nephrocystin-6, NPHP6centrosomal protein of 290 kDa, POC3, POC3 centriolar protein homolog, prostate cancer antigen T21, rd16,3H11AG, SLSN6nephrocytsin-6,3H11Ag, Tumor antigen se2-2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
CEP290 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:IIPSLERLVNAIESKNAEGIFDASLHLKAQVDQLTGRNEELRQELRESRKEAINYSQQLAKANLKIDHLEKETSLLRQSEGSNVVFKGIDLPDGIAPSSASII | |
25 μL | |
Vision | |
80184 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction